Anti-CENPA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-0483R-A488
BOSSBS-0483R-A488EA 1190 CHF
BOSSBS-0483R-A488
Anti-CENPA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatinMore Product Information