Anti-Digoxin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0356R-A750
BOSSBS-0356R-A750EA 1190 CHF
BOSSBS-0356R-A750
Anti-Digoxin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Digoxin is a beta blocker drug originally derived from the foxglove plant, Digitalis lanata. Digoxin is used primarily to improve the pumping ability of the heart in congestive heart failure (CHF), and treat problems such as high blood pressure. It is also used to help normalize some dysrhythmias (abnormal types of heartbeat).

Type: Primary
Antigen: Digoxin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR