Anti-GCH1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-0136R-A647
BOSSBS-0136R-A647EA 1190 CHF
BOSSBS-0136R-A647
Anti-GCH1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
GTP cyclohydrolase I (also designated dopa-responsive dystonia) catalyzes the conversion of GTP to D-erythro-7,8-dihydroneopterin triphosphate, the first and rate-limiting step in tetrahydrobiopterin (BH4) biosynthesis. Tetrahydrobiopterin is an essential cofactor for 3 aromatic amino acid monooxygenases: phenylalanine, tyrosine, and tryptophan hydroxylases. Animals can synthesize tetrahydrobiopterin in vivo from GTP through several enzymatic reactions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTP cyclohydrolase 1
Clonality: Polyclonal
Clone:
Conjugation: AlexaMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR