Anti-BAX Mouse Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-0127M-CY5
BOSSBS-0127M-CY5EA 1210 CHF
BOSSBS-0127M-CY5
Anti-BAX Mouse Polyclonal Antibody (Cy5®)
Antikörper
Accelerates programmed cell death by binding to, and antagonizing the apoptosis repressor BCL2 or its adenovirus homolog E1B 19k protein. Under stress conditions, undergoes a conformation change that causes translocation to the mitochondrion membrane, leading to the release of cytochrome c that then triggers apoptosis. Promotes activation of CASP3, and thereby apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BAX
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR