Tubing, PTFE, heat shrinkable, BOLA
Lieferant: Bohlender
S1828-64 S1828-32 S1828-40 S1828-48 S1828-16 S1828-24 S1828-56 S1828-08
BOHLS1828-64EA 75.4 CHF
BOHLS1828-64 BOHLS1828-32 BOHLS1828-40 BOHLS1828-48 BOHLS1828-16 BOHLS1828-24 BOHLS1828-56 BOHLS1828-08
Tubing, PTFE, heat shrinkable, BOLA
Schläuche Schrumpfschläuche
PTFE, translucent.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Shrink rate: 4:1
  • Incombustible (ASTM D876)
  • Insulation resistance 1018 O/cm (ASTM D 876)
  • Good electric strength
  • Flame retardant
For protection of probes, cables, electric components etc. against chemical disturbance. The shrink rate of 4:1 means that the inner diameter of the tubing shrinks to approx. ¼ of the original inner diameter and that the length shrinks to approx. 15%. Good heat transmission due to low wall thickness.