Tweezers, BOLA
Lieferant: Bohlender
H909-04 H912-02 H912-04 H912-06 H909-06 H909-02
BOHLH909-04EA 32 CHF
BOHLH909-04 BOHLH912-02 BOHLH912-04 BOHLH912-06 BOHLH909-06 BOHLH909-02
Tweezers, BOLA
Pinzetten
Precast tweezers made of PTFE.
  • With pointed or blunt ends
  • Universal chemical resistance
  • Temperature resistant from −200 to +250 °C
Order Now
TECHNISCHE DATEN
Environmentally Preferable