Double bossheads, swivel type
Lieferant: BOCHEM
5362
BOCH5362EA 41.9 CHF
BOCH5362
Double bossheads, swivel type
Klemmen Stativmuffen
Double bossheads made of malleable cast iron, chromium plated.
  • Swivel type
  • For rods up to 16,5 mm
  • Angle: 0 to 360°
  • With thumb screw (M8)
Order Now
TECHNISCHE DATEN
Environmentally Preferable