Anti-MVP Mouse Monoclonal Antibody (CF594) [clone: 1014]

Lieferant: Biotium
BNC940224-500 BNC940224-100
BNC940224-500EA 671 CHF
BNC940224-500 BNC940224-100
Anti-MVP Mouse Monoclonal Antibody (CF594) [clone: 1014]
Antikörper
Recognizes a protein of 104 kDa-110 kDa, characterized as major vault protein (MVP). Vaults are large ribonucleoprotein particles (RNPs) present in all eukaryotic cells. They have a complex morphology, including several small molecules of RNA, but a single protein species. The MVP accounts for >70% of their mass. Their shape is reminiscent of the nucleopore central plug. Treatment of cells with estradiol increases the amount of MVP in nuclear extract. The hormone-dependent interaction of vaults with ER is prevented in vitro by sodium molybdate. Antibodies to estrogen, progesterone andMore Product Information
Order Now

TECHNISCHE DATEN

Antigen-Symbol MVP
Environmentally Preferable
Antigen-Name Major Vault Protein
Antigen-Synonyme MVP|VAULT1|Major Vault Protein|Lung resistance-related protein (LRP)
Antikörper-Typ Primär
Klonalität Monoklonal
Konjugation CF® 594
Klon Clone 1014
Reaktivität Human
Wirt Mouse
Gen-ID 632177
Isotyp IgG1 kappa
ImmunFluoreszenz Yes
Konzentration 0,1 mg/ml
Lagerungspuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR