Anti-UGT1A9 Mouse Monoclonal Antibody (CF647) [clone: UGT1A9/1229]
Lieferant: Biotium
BNC471229-500 BNC471229-100
BNC471229-500EA 671 CHF
BNC471229-500 BNC471229-100
Anti-UGT1A9 Mouse Monoclonal Antibody (CF647) [clone: UGT1A9/1229]
Antikörper
This antibody recognizes a protein of about 60 kDa, which is identified as human UGT1A9. It does not cross-react with the other UGT1A isoforms including UGT1A7, UGT1A8, and UGT1A10 and shows a high degree of specificity. UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This MAb binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC s). In frozen sections, it stains hepatic cells and may be usedMore Product Information