Pipette tip reload, Rainin® LTS® pipette compatible xTIP4™ CleanPak™
Lieferant: BIOTIX
63300010 63300011 63300013 63300012 63300027
BITX63300010EA 72.9 CHF
BITX63300010 BITX63300011 BITX63300013 BITX63300012 BITX63300027
Pipette tip reload, Rainin® LTS® pipette compatible xTIP4™ CleanPak™
Pipettenspitzen
Create a perfect ergonomic system in the lab with FlexFit®, StarStop®, and a Biotix xPIPETTE or Rainin LTS® pipette.
  • Low cost option to get high-quality tips
  • 960 tips on the lab bench using the same surface area as 1 rack
  • Re-sealable bag is available in both sterile and nonsterile formats
  • Colour-coded snap cards allow for quick identification
  • The product is fully recyclable and contains no contaminating cardboard or paper
  • Inserts are easy to transfer to an Optimus empty rack
  • Full multi-channel and single-channel xPIPETTE and LTS® pipette compatibility