Anti-PERP Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB74448-100
BIRBORB74448-100EA 0 CHF
BIRBORB74448-100
Anti-PERP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PERP (PERP, TP53 apoptosis effector)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR