Anti-Maltose Binding Protein Mouse Monoclonal Antibody

Lieferant: Biorbyt
ORB26045-0.2
BIRBORB26045-0.2EA 0 CHF
BIRBORB26045-0.2
Anti-Maltose Binding Protein Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Maltose Binding Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR