Anti-Chloramphenicol Mouse Monoclonal Antibody

Lieferant: Biorbyt
ORB24471-1
BIRBORB24471-1EA 0 CHF
BIRBORB24471-1
Anti-Chloramphenicol Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Chloramphenicol
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR