Anti-SELL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)

Lieferant: Biorbyt
ORB129054-100
BIRBORB129054-100EA 0 CHF
BIRBORB129054-100
Anti-SELL Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Antikörper
Type: Primary
Antigen: SELL (selectin L)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR