Anti-KSR2 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB127012-100 ORB127012-200
BIRBORB127012-100EA 0 CHF
BIRBORB127012-100 BIRBORB127012-200
Anti-KSR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KSR2 (kinase suppressor of ras 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR