Anti-ATP1B2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Biorbyt
ORB122992-100
BIRBORB122992-100EA 0 CHF
BIRBORB122992-100
Anti-ATP1B2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Type: Primary
Antigen: ATP1B2 (ATPase, Na+/K+ transporting, beta 2 polypeptide)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR