Anti-TNFRSF8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Biorbyt
ORB110079-100
BIRBORB110079-100EA 0 CHF
BIRBORB110079-100
Anti-TNFRSF8 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR