Anti-GFP Rabbit polyclonal antibody

Lieferant: Biorbyt
ORB10708-100 ORB10708-200
BIRBORB10708-100EA 0 CHF
BIRBORB10708-100 BIRBORB10708-200
Anti-GFP Rabbit polyclonal antibody
Antikörper
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR