Cleanroom wipes, MicroPolx® 4000-HP
Lieferant: BERKSHIRE
MP4000HP090910
BERKMP4000HP090910EA 2110 CHF
BERKMP4000HP090910
Cleanroom wipes, MicroPolx® 4000-HP
Wischtücher Reinraum-Wischtücher Trockene Reinraumwischtücher
These wipes made from 80% Polyester and 20% nylon micro denier knitted fabric are designed for the highest level of contamination control in critical processing applications.
  • Microfibre yarn increases surface area and provides superior wiping efficiency and sorptive performance
  • Cleans efficiently with less solvent usage than traditional wipes
  • Removes oily soils with little or no solvent required
  • Soft hand and non-abrasive sealed-edge are excellent for sensitive or scratch prone surfaces
  • Ultrasonically sealed edges minimise fibre and particle generation
More Product Information