Digital saccharimeters, ADS400 series
Lieferant: BELLINGHAM STANLEY
37-25 37-26 37-27 37-85 37-86 37-87
BELL37-25EA 10450 CHF
BELL37-25 BELL37-26 BELL37-27 BELL37-85 BELL37-86 BELL37-87
Digital saccharimeters, ADS400 series
Polarimeter
Specifically suited for use in sugar processing industry. Rapid analysis of POL and purity of raw or processed sugar in the laboratory or Tare House.
  • Single wavelength (589 or NIR 850 nm)
  • Standard or funnel-flow tubes
  • On-board purity calculation
  • Multi-lingual display
  • Continuous reading
ADS400 Series Saccharimeters are specifically designed to meet the requirements of users wishing to measure optical rotation of sucrose solutions within the sugar processing industry.
A choice of instrument type facilitates a multitude of configurations, the mainMore Product Information