Screw caps with liners, for culture tubes and flasks
Lieferant: BELLCO GLASS
2029-00016 2029-00051 5636-33430 2029-00038 5636-38430
BELC2029-00016EA 53.91 CHF
BELC2029-00016 BELC2029-00051 BELC5636-33430 BELC2029-00038 BELC5636-38430
Screw caps with liners, for culture tubes and flasks
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
Screw caps are made from mineral-filled phenolic resin to resist the effects of temperature and steam autoclaving. They have a non-toxic white rubber liner for all general culture applications.
  • Short skirt for use with glass roller bottles
  • Deep skirt reduces risk of contamination