Photo-etched cover glasses
Lieferant: BELLCO GLASS
1916-92525 1916-91818 1916-91025 1916-91012
BELC1916-92525EA 72.76 CHF
BELC1916-92525 BELC1916-91818 BELC1916-91025 BELC1916-91012
Photo-etched cover glasses
Deckgläser
Cover glasses used for fluorescence and confocal microscopy.