Reagent reservoirs, 12 channel, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F378801200
BELAF378801200EA 110 CHF
BELAF378801200
Reagent reservoirs, 12 channel, SP Bel-Art
Reagenz-Reservoirs
PP. This chemical resistant 12 channel reagent reservoir can be used with single, 8 and 12 channel pipettes.
  • Wide profile provides added stability
  • Autoclavable at 121 °C
  • Individually sealed and sterilized (8 per bag, 6 bags per case)
  • V-shaped wells allow complete sample withdrawal
  • Each channel is numbered to facilitate serial dilution and holds 5 ml
  • Fitted lid included