Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F378760002
BELAF378760002EA 60.7 CHF
BELAF378760002
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Mikroplatten- und Petrischalen-Replikatoren
The 96 'open-ended' pipette tips in this replicating tool are compatible with all kinds of plates and tubes.
  • Fits plates with flat, V- or U-bottom wells
  • Up to 10 µl of fluid per tip aspirated
  • Sturdy, durable polypropylene construction
  • Re-usable and steam sterilisable