Sampling scoops, Behrotest®
Lieferant: BEHR
B00196081 B00196079 B00196080
BEHRB00196081EA 790 CHF
BEHRB00196081 331-3110 331-3120
Sampling scoops, Behrotest®
Probenehmer Probenehmer für Wasser
The sampling scoop is composed of a clamping fixture and end bar. The scoop with receptacles made of borosilicate glass comply with the high requirements.
  • Sturdy three-part supporting bar with telescopic racks
  • Continuously adjustable in length
Suitable for sampling in waters that are difficult to access, from the surface.
Zertifizierungen: EN 1485/AOX
Order Now
TECHNISCHE DATEN
Environmentally Preferable