Locator™ 6 Rack and Box System
Lieferant: Thermo Fisher Scientific
CY50985 CY509113
478-0808EA 9590 CHF
478-0808 BARNCY509113
Locator™ 6 Rack and Box System
Dewars und Kryogefäße
Economically store up to 6000 samples in this cryogenic storage system that makes sample retrieval simple and consumes minimum liquid nitrogen.
  • Compact design minimizes space requirements while maximizing storage capacity.
  • System is suited for both manual and computerized inventory record-keeping methods.
  • Samples are stored below –180 °C, even when less than 2 in. (5 cm) of liquid nitrogen remains in the vessel.
  • Optional Ultrasonic Level Monitor safeguards irreplaceable samples with minimal liquid nitrogen evaporation and conduction.
  • Audible alarm sounds
More Product Information