Incubating orbital shakers, MaxQ™ HP
Lieferant: Thermo Fisher Scientific
238000 238001 238008 238002
BARN238000EA 2870 CHF
BARN238000 BARN238001 BARN238008 BARN238002
Incubating orbital shakers, MaxQ™ HP
Schüttler Inkubationsschüttler
Max Q™ HP combines incubated or refrigerated temperature with an orbital shaker.
  • Triple counter-balanced mechanism
  • Microprocessor functions are calibrated through keypad
  • Clear acrylic lid withstands years of use without cracking or discoloration
  • Audible over and under-temperature alarms ensure sample protection
  • Innovative design ensures system protection in case of spills
Features a double walled insulated chamber which ensures temperature uniformity of ±0,3 °C. The spacious stainless steel chamber allows stacking of a 457×457 mm platform to hold six 2More Product Information