L(-)-Tryptophan

Lieferant: BACHEM BIOCHEMICA

Synonyms: L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH

4031215.0100 4031215.0250 4031215.1000
BACE4031215.0100EA 0 CHF
BACE4031215.0100 BACE4031215.0250 BACE4031215.1000
L(-)-Tryptophan
L(-)-Tryptophan
Formel: C₁₁H₁₂N₂O₂
Melting Pt: 290 °C
Dichte: 1.34 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 12,09929

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR