L-Methioninsulfoxid

Lieferant: BACHEM BIOCHEMICA

Synonyms: H-Met(O)-OH, (2S)-2-Amino-4-(methylsulphinyl)butanoic acid

4014135.0005 4014135.0025
BACE4014135.0005EA 0 CHF
BACE4014135.0005 BACE4014135.0025
L-Methioninsulfoxid
L-Methioninsulfoxid
Formel: C₅H₁₁NO₃S
MW: 165.2 g/mol
MDL Number: MFCD00063093
CAS-Nummer: 3226-65-1
EINECS: 221-758-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR