4-Iod-D-phenylalanin

Lieferant: BACHEM BIOCHEMICA

Synonyms: H-D-Phe(4-I)-OH

4010046.0250
BACE4010046.0250EA 0 CHF
BACE4010046.0250
4-Iod-D-phenylalanin
4-Iod-D-phenylalanin
Formel: C₉H₁₀INO₂
MDL Number: MFCD00063067
CAS-Nummer: 62561-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR