Toluol-α-sulfonylfluorid

Lieferant: BACHEM BIOCHEMICA
Gefahr

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

4000472.0025 4000472.0100
BACE4000472.0025EA 0 CHF
BACE4000472.0025 BACE4000472.0100
Toluol-α-sulfonylfluorid
Toluol-α-sulfonylfluorid
Formel: C₇H₇FO₂S
Siedepunkt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007424
CAS-Nummer: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR