2-Aminoterephthalsäure 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Aminoterephthalic Acid

B24441.06 B24441.14 B24441.22
B24441.06EA 59.9 CHF
B24441.06 B24441.14 B24441.22
2-Aminoterephthalsäure 99%
2-Aminoterephthalsäure

Purity: 99% CAS No.: 10312-55-7 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odour: Odourless Melting Point: 300° MDL No.: MFCD00134536

Formel: C₈H₇NO₄
MW: 181.15 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00134536
CAS-Nummer: 10312-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR