1-(Dimethylamino)-2-propanol 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: N,N-Dimethyl-1-amino-2-propanol, Dimepranol, N,N-Dimethylisopropanolamine

B24234.0B B24234.30
B24234.0BEA 134 CHF
B24234.0B B24234.30
1-(Dimethylamino)-2-propanol 99%
1-(Dimethylamino)-2-propanol

Purity: 99% CAS No.: 108-16-7 Molecular Formula: C5H13NO Formula Weight: 103.16 Flash Point: 35°(95°F) Refractive Index: 1.4195 Odour: Amine-like pH: 11 Melting Point: -85° Boiling Point: 123-126° UN No.: 2734 MDL No.: MFCD00004532

Formel: C₅H₁₃NO
Siedepunkt: 123…126 °C
Melting Pt: –85 °C
Dichte: 0.85
Flash-Pt: 35 °C (95 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004532
CAS-Nummer: 108-16-7
EINECS: 203-556-4
UN: 2734
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR