Saccharin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyms: o-Sulphobenzimide, o-Benzoic sulphimide

B23938.0E B23938.22 B23938.36
B23938.0EEA 708 CHF
B23938.0E B23938.22 B23938.36
Saccharin ≥98%
Saccharin

Purity: 98+% CAS No.: 81-07-2 Molecular Formula: C7H5NO3S Formula Weight: 183.18 Melting Point: 227-230° Beilstein Reference: 6888 Merck Reference: 14,8311 MDL No.: MFCD00005866

Formel: C₇H₅NO₃S
MW: 183.19 g/mol
Melting Pt: 227…230 °C
Dichte: 0.828
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005866
CAS-Nummer: 81-07-2
EINECS: 201-321-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR