3-Amino-1-propanol 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 3-Amino-1-propanol

B23041.22 B23041.36
B23041.22EA 58.4 CHF
B23041.22 B23041.36
3-Amino-1-propanol 99%
3-Amino-1-propanol

Purity: 99% CAS No.: 156-87-6 Molecular Formula: H2N(CH2)3OH Formula Weight: 75.11 Flash Point: 101°(214°F) Refractive Index: 1.4610 Odour: Amine-like Melting Point: 11° Boiling Point: 184-187° Beilstein Reference: 741855 UN No.: 2735 MDL No.: MFCD00008223

Formel: C₃H₉NO
Siedepunkt: 184…187 °C
Melting Pt: 11 °C
Dichte: 0.983
Flash-Pt: 101 °C (214 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008223
CAS-Nummer: 156-87-6
EINECS: 205-864-4
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR