4-Isopropylphenol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B22620.18 B22620.30 B22620.A1
B22620.18EA 64.3 CHF
B22620.18 B22620.30 B22620.A1
4-Isopropylphenol ≥98%
4-Isopropylphenol

Purity: 98% CAS No.: 99-89-8 Molecular Formula: C9H12O Formula Weight: 136.20 Flash Point: 108°(226°F) Melting Point: 59-61° Boiling Point: 211-212° Beilstein Reference: 1363564 UN No.: 2430 MDL No.: MFCD00002372

Formel: C₉H₁₂O
Siedepunkt: 211…212 °C
Melting Pt: 59…61 °C
Dichte: 0.99 g/cm³ (20 °C)
Flash-Pt: 108 °C (226 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002372
CAS-Nummer: 99-89-8
EINECS: 202-798-8
UN: 2430
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR