2-Fluor-α-methyl-4-biphenylessigsäure ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

B22603.03 B22603.06 B22603.14
B22603.03EA 135 CHF
B22603.03 B22603.06 B22603.14
2-Fluor-α-methyl-4-biphenylessigsäure ≥99%
2-Fluor-α-methyl-4-biphenylessigsäure

Purity: 99% CAS No.: 5104-49-4 Molecular Formula: C15H13FO2 Formula Weight: 244.27 Odour: Odourless Melting Point: 112-114° UN No.: 2811 MDL No.: MFCD00079303

Formel: C₁₅H₁₃FO₂
MW: 244.265 g/mol
Melting Pt: 112…114 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6
ADR: 6.1 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR