5-Chlor-1,3-benzodioxol 98

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 5-Chlorobenzo[d][1,3]dioxole

B22534.09 B22534.18
B22534.09EA 77.8 CHF
B22534.09 B22534.18
5-Chlor-1,3-benzodioxol 98
5-Chlor-1,3-benzodioxol

Purity: 98% CAS No.: 7228-38-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Flash Point: 93°(201°F) Refractive Index: 1.5560 Boiling Point: 185-187° MDL No.: MFCD00010842

Formel: C₇H₅ClO₂
MW: 156.57 g/mol
Siedepunkt: 185…187 °C
Dichte: 1.340
Flash-Pt: 93 °C (201 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010842
CAS-Nummer: 7228-38-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR