4-Butylbenzylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: (4-Butylphenyl)methanamine

B22316.06 B22316.14
B22316.06EA 175 CHF
B22316.06 B22316.14
4-Butylbenzylamin 98%
4-Butylbenzylamin

Purity: 98% CAS No.: 57802-79-6 Molecular Formula: C11H17N Formula Weight: 163.26 Boiling Point: 89-91°/1mm UN No.: 2735 MDL No.: MFCD02258855

Formel: C₁₁H₁₇N
MW: 163.26 g/mol
Siedepunkt: 89…91 °C (1 mmHg)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02258855
CAS-Nummer: 57802-79-6
ADR: 8 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR