Säurefuchsin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)

B22222.14 B22222.22 B22222.36
B22222.14EA 145 CHF
B22222.14 B22222.22 B22222.36
Säurefuchsin
Säurefuchsin

CAS No.: 3244-88-0 Molecular Formula: C20H17N3Na2O9S3 Formula Weight: 585.53 Beilstein Reference: 4111656 Merck Reference: 14,107 MDL No.: MFCD00013286

Formel: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Melting Pt: > 130 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013286
CAS-Nummer: 3244-88-0
EINECS: 221-816-5
Merck Index: 14,107

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR