Tolbutamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: N-(Butylcarbamoyl)-4-methylbenzenesulphonamide

B21698.09 B21698.18 B21698.30
B21698.09EA 76.9 CHF
B21698.09 B21698.18 B21698.30
Tolbutamid ≥98%
Tolbutamid

Purity: 98% CAS No.: 64-77-7 Molecular Formula: C12H18N2O3S Formula Weight: 270.35 Melting Point: 128-130° Beilstein Reference: 1984428 Merck Reference: 14,9507 MDL No.: MFCD00027169

Formel: C₁₂H₁₈N₂O₃S
Melting Pt: 128…130 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00027169
CAS-Nummer: 64-77-7
EINECS: 200-594-3
Merck Index: 14,9507

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR