Safranin O

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Gossypimine, Safranin O, Safranin Y, Safranin A, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, 3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chloride, Safranin T, CI 50240, Basic Red 2, Safranin, Cotton Red

B21674.09 B21674.18
B21674.09EA 50.9 CHF
B21674.09 B21674.18
Safranin O
Safranin O

CAS No.: 477-73-6 Molecular Formula: C20H19ClN4 Formula Weight: 350.85 Odour: Odourless Beilstein Reference: 3924099 MDL No.: MFCD00011759

Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Dichte: 1.447 g/cm³ (22 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011759
CAS-Nummer: 477-73-6
EINECS: 207-518-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR