Chlorbenzol 99

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

B21052.0F B21052.AP
B21052.0FEA 135 CHF
B21052.0F B21052.AP
Chlorbenzol 99
Chlorbenzol

Purity: 99% CAS No.: 108-90-7 Molecular Formula: C6H5Cl Formula Weight: 112.56 Flash Point: 28°(82°F) Refractive Index: 1.5240 Odour: Aromatic Melting Point: -45° Boiling Point: 132° Beilstein Reference: 605632 Merck Reference: 14,2121 UN No.: 1134 MDL No.: MFCD00000530

Formel: C₆H₅Cl
MW: 112.56 g/mol
Siedepunkt: 132 °C
Melting Pt: –45 °C
Dichte: 1.106
Flash-Pt: 28 °C (82 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000530
CAS-Nummer: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3 III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR