5-Fluorisatin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 5-Fluoroindoline-2,3-dione, 5-Fluoro-1H-indole-2,3-dione

B20831.03 B20831.06 B20831.14
B20831.03EA 43.1 CHF
B20831.03 B20831.06 B20831.14
5-Fluorisatin ≥98%
5-Fluorisatin

Purity: 98% CAS No.: 443-69-6 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 224-227° Beilstein Reference: 131241

Formel: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 224…227 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00022795
CAS-Nummer: 443-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR