Anti-OGFOD1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP48825_P050
AVIVARP48825_P050EA 533 CHF
AVIVARP48825_P050
Anti-OGFOD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OGFOD1 (2-oxoglutarate and iron-dependent oxygenase domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR