Anti-ACMSD Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP45728_P050
AVIVARP45728_P050EA 533 CHF
AVIVARP45728_P050
Anti-ACMSD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACMSD (aminocarboxymuconate semialdehyde decarboxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR