Anti-AGER Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP41464_P050
AVIVARP41464_P050EA 533 CHF
AVIVARP41464_P050
Anti-AGER Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR