Natriumhypochlorit solution_SC 13% w/v, technical

Lieferant: PanReac AppliChem

213322.1214 213322.0715
APPC213322.1214EA 82.6 CHF
APPC213322.1214 APPC213322.0715
Natriumhypochlorit solution_SC 13% w/v, technical
Sodium hypochlorite
Formel: ClNaO
MW: 74.44 g/mol
Melting Pt: -6
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011120
CAS-Nummer: 7681-52-9
UN: 1791
ADR: 8 III
Merck Index: 13,08702

Order Now

Spezifikation Testergebnisse

Assay (Active Chlorine, Iodometry) 10.0 - 14.0%
NaCl Max. 11.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR