Natriumhydroxid -Lösung 40% (w/w)

Lieferant: PanReac AppliChem
Gefahr

Synonyms: Caustic soda

171220.1214 171220.1315 171220.0716 171220.0715
APPC171220.1214EA 488 CHF
APPC171220.1214 APPC171220.1315 APPC171220.0716 APPC171220.0715
Natriumhydroxid -Lösung 40% (w/w)
Sodium hydroxide

Absorbs CO₂ from air. Miscible with water.

Formel: NaOH
Dichte: 1.43 g/ml
MDL Number: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Spezifikation Testergebnisse

Minimum Assay (Acidim.) 40%
Nitrogen Compounds (as N) Max. 0.002%
Carbonate (as Na₂CO₃) Max. 1.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR