Magnesiumchlorid Hexahydrat 98,0-101,0 Ph. Eur., BP, pharma grade

Lieferant: PanReac AppliChem

Synonyms: Magnesium dichloride

141396.0416 141396.0914 141396.1211
APPC141396.0416EA 0 CHF
APPC141396.0416 APPC141396.0914 APPC141396.1211
Magnesiumchlorid Hexahydrat 98,0-101,0 Ph. Eur., BP, pharma grade
Magnesiumchlorid Hexahydrat
Formel: MgCl₂·6H₂O
MW: 203.3 g/mol
Siedepunkt: 1412
Melting Pt: 117 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149781
CAS-Nummer: 7791-18-6
Merck Index: 13,05684

Order Now

Spezifikation Testergebnisse

Assay (Compl.) 98.0 - 101.0%
Identity according to Pharmacopoeias Passes Test
Appearance of Solution Passes Test
Acidity or Alkalinity Passes Test
Ammonium (NH₄) 0.005%
Bromide (Br) 0.05%
Phosphate (PO₄) 0.003%
Sulfate (SO₄) 0.01%
Residual solvents (Ph.Eur.) Passes Test
Water (H₂O) 51.0 - 55.0%
Heavy metals (as Pb) 0.001%
Al 0.0001%
As 0.00015%
Ca 0.1%
Fe 0.001%
Cd, (Class 1 - ICH Q3D) 0,5 ppm
Pb, (Class 1 - ICH Q3D) 0,5 ppm
As, (Class 1 - ICH Q3D) 1,5 ppm
Hg, (Class 1 - ICH Q3D) 1,5 ppm
Co, (Class 2A - ICH Q3D) 5 ppm
V, (Class 2A - ICH Q3D) 10 ppm
Ni, (Class 2A - ICH Q3D) 20 ppm
Tl, (Class 2B - ICH Q3D) 0,8 ppm
Au, (Class 2B - ICH Q3D) 10 ppm
Pd, (Class 2B - ICH Q3D) 10 ppm
Ir, (Class 2B - ICH Q3D) 10 ppm
Os, (Class 2B - ICH Q3D) 10 ppm
Rh, (Class 2B - ICH Q3D) 10 ppm
Ru, (Class 2B - ICH Q3D) 10 ppm
Se, (Class 2B - ICH Q3D) 15 ppm
Ag, (Class 2B - ICH Q3D) 15 ppm
Pt, (Class 2B - ICH Q3D) 10 ppm
Li, (Class 3 - ICH Q3D) 55 ppm
Sb, (Class 3 - ICH Q3D) 120 ppm
Ba, (Class 3 - ICH Q3D) 140 ppm
Mo, (Class 3 - ICH Q3D) 25 ppm
Cu, (Class 3 - ICH Q3D) 250 ppm
Sn, (Class 3 - ICH Q3D) 600 ppm
Cr, (Class 3 - ICH Q3D) 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR